siRNA Details
| Gene Symbol: Snx16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022289.3 | NP_071625.2 | 2 | 425 | ||
| XM_006232135.2 | XP_006232197.1 | 2 | 438 | |||
| GenBank mRNA | AF068746.2 | - | 1 | 137 | ||
| AF305780.1 | - | 2 | 411 | |||
| BC061554.1 | - | 2 | 429 | 6890629 | ||

Snx16
Genomic Map