siRNA Details
| Gene Symbol: Mlst8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022404.2 | NP_071799.2 | 3, 4 | 550 | ||
| GenBank mRNA | AF051155.2 | - | 3, 4 | 549 | ||
| BC097319.1 | - | 3, 4 | 228 | 7455095 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mlst8| Gene Symbol: Mlst8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022404.2 | NP_071799.2 | 3, 4 | 550 | ||
| GenBank mRNA | AF051155.2 | - | 3, 4 | 549 | ||
| BC097319.1 | - | 3, 4 | 228 | 7455095 | ||