siRNA Details
| Gene Symbol: Casp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022522.2 | NP_071967.2 | 1, 2 | 88 | ||
| GenBank mRNA | AF136231.1 | - | 1, 2 | 86 | ||
| AF136232.1 | - | 1, 2 | 86 | |||
| U77933.1 | - | 1, 2 | 88 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Casp2| Gene Symbol: Casp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022522.2 | NP_071967.2 | 1, 2 | 88 | ||
| GenBank mRNA | AF136231.1 | - | 1, 2 | 86 | ||
| AF136232.1 | - | 1, 2 | 86 | |||
| U77933.1 | - | 1, 2 | 88 | |||