siRNA Details
Gene Symbol: Tcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022534.1 | NP_071979.1 | 3 | 323 | ||
XM_006251366.2 | XP_006251428.1 | 4 | 514 | |||
XM_006251367.2 | XP_006251429.1 | 4 | 508 | |||
XM_006251368.2 | XP_006251430.1 | 4 | 453 | |||
XM_006251370.2 | XP_006251432.1 | 4 | 446 | |||
GenBank mRNA | AF054810.1 | - | 3 | 323 |
