siRNA Details
| Gene Symbol: Entpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022587.1 | NP_072109.1 | 2 | 196 | ||
| GenBank mRNA | U81295.1 | - | 2 | 196 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entpd1| Gene Symbol: Entpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022587.1 | NP_072109.1 | 2 | 196 | ||
| GenBank mRNA | U81295.1 | - | 2 | 196 | ||