siRNA Details
| Gene Symbol: Entpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022587.1 | NP_072109.1 | 7 | 1004 | ||
| GenBank mRNA | U81295.1 | - | 7 | 1004 | ||
| Gene Symbol: LOC103689954 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760464.1 | XP_008758686.1 | 940 | |||

Entpd1
Genomic Map