siRNA Details
| Gene Symbol: Rem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022685.2 | NP_073176.2 | 5 | 857 | ||
| GenBank mRNA | AF084464.1 | - | 5 | 731 | ||
| AY916790.1 | - | 5 | 797 | |||
| BC127495.1 | - | 5 | 842 | 8371030 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rem2| Gene Symbol: Rem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022685.2 | NP_073176.2 | 5 | 857 | ||
| GenBank mRNA | AF084464.1 | - | 5 | 731 | ||
| AY916790.1 | - | 5 | 797 | |||
| BC127495.1 | - | 5 | 842 | 8371030 | ||