siRNA Details
| Gene Symbol: Arl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022700.2 | NP_073191.1 | 1 | 124 | ||
| XM_006231559.2 | XP_006231621.1 | 2 | 645 | |||
| GenBank mRNA | BC084722.1 | - | 1 | 124 | 7310623 | |
| U12568.1 | - | 1 | 128 | |||
| X76921.1 | - | 2 | 256 | |||

Arl3
Genomic Map