siRNA Details
| Gene Symbol: Gabarapl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022706.2 | NP_073197.1 | 4 | 355 | ||
| GenBank mRNA | AB003515.1 | - | 4 | 393 | ||
| BC058145.1 | - | 4 | 358 | 6921109 | ||
| BC088139.1 | - | 4 | 355 | 7323806 | ||
| FQ216852.1 | - | 4 | 369 | |||
| FQ226397.1 | - | 4 | 379 | |||
| FQ231643.1 | - | 4 | 348 | |||
| FQ233551.1 | - | 4 | 390 | |||
| FQ234861.1 | - | 4 | 348 | |||
| Gene Symbol: LOC100359937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729947.4 | XP_002729993.3 | 1 | 344 | ||
| Gene Symbol: LOC103692360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763731.1 | XP_008761953.1 | 5 | 725 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_026693, BC081436, AK049819, AK134873, AK002596, and others.

Gabarapl2
Genomic Map