siRNA Details
Gene Symbol: Cplx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022864.3 | NP_074055.1 | 3 | 1028 | ||
XM_006250571.2 | XP_006250633.1 | 4 | 1154 | |||
GenBank mRNA | BC093605.1 | - | 3 | 1031 | 7317504 | |
D70817.1 | - | 4 | 1063 |
