siRNA Details
| Gene Symbol: Gphn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022865.3 | NP_074056.2 | 3 | 314 | ||
| GenBank mRNA | X66366.2 | - | 3 | 444 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gphn| Gene Symbol: Gphn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022865.3 | NP_074056.2 | 3 | 314 | ||
| GenBank mRNA | X66366.2 | - | 3 | 444 | ||