siRNA Details
| Gene Symbol: Kcnip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001261387.1 | NP_001248316.1 | 4 | 430 | ||
| NM_001261388.1 | NP_001248317.1 | 3 | 397 | |||
| NM_001261389.1 | NP_001248318.1 | 4 | 266 | |||
| NM_022929.2 | NP_075218.2 | 3 | 233 | |||
| XM_006246105.2 | XP_006246167.1 | 3 | 1288 | |||
| GenBank mRNA | AB046443.1 | - | 3 | 233 | ||
| AY082657.1 | - | 4 | 266 | |||
| AY082658.1 | - | 3 | 233 | |||
| AY142709.1 | - | 4 | 266 | |||

Kcnip1
Genomic Map