siRNA Details
| Gene Symbol: Htr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231307.2 | XP_006231369.1 | 3 | 2454 | ||
| GenBank mRNA | L22558.1 | - | 3 | 2239 | ||
| X69663.1 | - | 3 | 2407 | |||
| Gene Symbol: LOC103694905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773900.1 | XP_008772122.1 | 2454 | |||
| XM_008773901.1 | XP_008772123.1 | 2459 | ||||
More Information
Set Membership: |
|

Htr7
Genomic Map