siRNA Details
| Gene Symbol: Clpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022947.1 | NP_075236.1 | 2, 3 | 508 | ||
| XM_008759796.1 | XP_008758018.1 | 2, 3 | 772 | |||
| GenBank mRNA | AB027570.1 | - | 2, 3 | 508 | ||
| BC070917.1 | - | 2, 3 | 471 | 7105142 | ||

Clpb
Genomic Map