siRNA Details
| Gene Symbol: Slit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022953.2 | NP_075242.1 | 3 | 452 | ||
| GenBank mRNA | AB011530.1 | - | 3 | 452 | ||
| AB073213.1 | - | 3 | 371 | |||
| AB073214.1 | - | 3 | 377 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slit1| Gene Symbol: Slit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022953.2 | NP_075242.1 | 3 | 452 | ||
| GenBank mRNA | AB011530.1 | - | 3 | 452 | ||
| AB073213.1 | - | 3 | 371 | |||
| AB073214.1 | - | 3 | 377 | |||