siRNA Details
Gene Symbol: Rhag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023022.2 | NP_075411.2 | 10 | 1866 | ||
GenBank mRNA | AF531097.1 | - | 10 | 1866 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rhag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023022.2 | NP_075411.2 | 10 | 1866 | ||
GenBank mRNA | AF531097.1 | - | 10 | 1866 |