siRNA Details
| Gene Symbol: Dync2h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023024.1 | NP_075413.1 | 2, 3 | 595 | ||
| GenBank mRNA | AB041881.1 | - | 2, 3 | 595 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dync2h1| Gene Symbol: Dync2h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023024.1 | NP_075413.1 | 2, 3 | 595 | ||
| GenBank mRNA | AB041881.1 | - | 2, 3 | 595 | ||