siRNA Details
| Gene Symbol: Gabre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023091.1 | NP_075579.1 | 2 | 170 | ||
| GenBank mRNA | AF189262.1 | - | 2 | 170 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabre| Gene Symbol: Gabre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023091.1 | NP_075579.1 | 2 | 170 | ||
| GenBank mRNA | AF189262.1 | - | 2 | 170 | ||