siRNA Details
| Gene Symbol: Atp5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023093.1 | NP_075581.1 | 3, 4 | 366 | ||
| GenBank mRNA | BC061830.1 | - | 3, 4 | 366 | 5599635 | |
| FQ214031.1 | - | 2, 3 | 208 | |||
| FQ214146.1 | - | 2, 3 | 208 | |||
| J05266.1 | - | 3, 4 | 287 | |||

Atp5a1
Genomic Map