siRNA Details
| Gene Symbol: Atp5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023093.1 | NP_075581.1 | 12 | 1781 | ||
| GenBank mRNA | BC061830.1 | - | 12 | 1781 | 5599635 | |
| FQ214146.1 | - | 11 | 1623 | |||
| FQ225416.1 | - | 7 | 1096 | |||
| FQ225735.1 | - | 7 | 1096 | |||
| FQ227531.1 | - | 7 | 1096 | |||
| FQ227720.1 | - | 6 | 916 | |||
| J05266.1 | - | 1702 | ||||
| X56133.1 | - | 5 | 778 | |||

Atp5a1
Genomic Map