siRNA Details
| Gene Symbol: Gbgt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB081649.1 | - | 5, 6 | 293 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gbgt1| Gene Symbol: Gbgt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB081649.1 | - | 5, 6 | 293 | ||