siRNA Details
| Gene Symbol: Trpv4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023970.1 | NP_076460.1 | 11 | 1903 | ||
| XM_006249466.2 | XP_006249528.1 | 11 | 1932 | |||
| GenBank mRNA | AF263521.1 | - | 11 | 1903 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trpv4| Gene Symbol: Trpv4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023970.1 | NP_076460.1 | 11 | 1903 | ||
| XM_006249466.2 | XP_006249528.1 | 11 | 1932 | |||
| GenBank mRNA | AF263521.1 | - | 11 | 1903 | ||