siRNA Details
| Gene Symbol: Nrgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024140.2 | NP_077054.1 | 4 | 1038 | ||
| GenBank mRNA | L09119.1 | - | 4 | 1038 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrgn| Gene Symbol: Nrgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024140.2 | NP_077054.1 | 4 | 1038 | ||
| GenBank mRNA | L09119.1 | - | 4 | 1038 | ||