siRNA Details
| Gene Symbol: Apex1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024148.1 | NP_077062.1 | 4 | 367 | ||
| XM_006251913.1 | XP_006251975.1 | 4 | 408 | |||
| GenBank mRNA | AF309114.1 | - | 3 | 283 | ||
| AF311054.1 | - | 3 | 245 | |||
| BC078816.1 | - | 4 | 375 | 7127205 | ||
| D44495.1 | - | 4 | 367 | |||
| FQ220521.1 | - | 4 | 369 | |||
| L27076.1 | - | 4 | 351 | |||

Apex1
Genomic Map