siRNA Details
| Gene Symbol: Ncs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024366.1 | NP_077342.1 | 3, 4 | 296 | ||
| XM_006233925.2 | XP_006233987.1 | 3, 4 | 600 | |||
| XM_008761679.1 | XP_008759901.1 | 2, 3 | 191 | |||
| GenBank mRNA | L27421.1 | - | 3, 4 | 236 | ||
| X82188.1 | - | 3, 4 | 296 | |||

Ncs1
Genomic Map