siRNA Details
| Gene Symbol: Chrna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024485.1 | NP_077811.1 | 3, 4 | 303 | ||
| GenBank mRNA | X74832.1 | - | 3, 4 | 303 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrna1| Gene Symbol: Chrna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024485.1 | NP_077811.1 | 3, 4 | 303 | ||
| GenBank mRNA | X74832.1 | - | 3, 4 | 303 | ||