siRNA Details
| Gene Symbol: Grpel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024487.4 | NP_077813.1 | 3 | 259 | ||
| XM_006251142.2 | XP_006251204.1 | 4 | 452 | |||
| GenBank mRNA | BC097312.1 | - | 3 | 257 | 7387647 | |
| FQ219742.1 | - | 4 | 440 | |||
| FQ220295.1 | - | 3 | 250 | |||
| U62940.1 | - | 3 | 260 | |||

Grpel1
Genomic Map