siRNA Details
| Gene Symbol: Sost | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030584.1 | NP_085073.1 | 1 | 241 | ||
| GenBank mRNA | AF326741.1 | - | 1 | 241 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sost| Gene Symbol: Sost | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030584.1 | NP_085073.1 | 1 | 241 | ||
| GenBank mRNA | AF326741.1 | - | 1 | 241 | ||