siRNA Details
| Gene Symbol: Gpr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037295.1 | NP_001032372.1 | 2 | 1329 | ||
| NM_030831.1 | NP_110458.1 | 1 | 1499 | |||
| GenBank mRNA | BC127447.1 | - | 2 | 1257 | 8363428 | |
| U12184.1 | - | 1 | 1499 | |||
| X61496.1 | - | 2 | 1327 | |||
More Information
Set Membership: |
|

Gpr12
Genomic Map