siRNA Details
| Gene Symbol: Slc26a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030840.1 | NP_110467.1 | 4 | 489 | ||
| GenBank mRNA | AJ303372.1 | - | 4 | 489 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc26a5| Gene Symbol: Slc26a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030840.1 | NP_110467.1 | 4 | 489 | ||
| GenBank mRNA | AJ303372.1 | - | 4 | 489 | ||