siRNA Details
Gene Symbol: Slc26a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030840.1 | NP_110467.1 | 18 | 2074 | ||
GenBank mRNA | AJ303372.1 | - | 18 | 2074 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc26a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030840.1 | NP_110467.1 | 18 | 2074 | ||
GenBank mRNA | AJ303372.1 | - | 18 | 2074 |