siRNA Details
| Gene Symbol: Bmpr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030849.1 | NP_110476.1 | 12 | 1817 | ||
| GenBank mRNA | D38082.1 | - | 11 | 1674 | ||
| S75359.1 | - | 12 | 1817 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmpr1a| Gene Symbol: Bmpr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030849.1 | NP_110476.1 | 12 | 1817 | ||
| GenBank mRNA | D38082.1 | - | 11 | 1674 | ||
| S75359.1 | - | 12 | 1817 | |||