siRNA Details
| Gene Symbol: Myoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030865.1 | NP_110492.1 | 2, 3 | 799 | ||
| GenBank mRNA | AB019393.1 | - | 2, 3 | 795 | ||
| AF093567.1 | - | 2, 3 | 799 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myoc| Gene Symbol: Myoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030865.1 | NP_110492.1 | 2, 3 | 799 | ||
| GenBank mRNA | AB019393.1 | - | 2, 3 | 795 | ||
| AF093567.1 | - | 2, 3 | 799 | |||