siRNA Details
| Gene Symbol: Inpp4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031002.3 | NP_112264.3 | 26 | 4077 | ||
| GenBank mRNA | CB761394.1 | - | 1 | 194 | ||
| CO395558.1 | - | 1 | 65 | |||
| U26397.1 | - | 26 | 4077 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Inpp4a| Gene Symbol: Inpp4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031002.3 | NP_112264.3 | 26 | 4077 | ||
| GenBank mRNA | CB761394.1 | - | 1 | 194 | ||
| CO395558.1 | - | 1 | 65 | |||
| U26397.1 | - | 26 | 4077 | |||