siRNA Details
| Gene Symbol: Gmfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031032.2 | NP_112294.1 | 4, 5 | 281 | ||
| GenBank mRNA | BC081778.1 | - | 4, 5 | 281 | 7192655 | |
| FQ222145.1 | - | 4, 5 | 343 | |||
| Z11558.1 | - | 3, 4 | 201 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gmfb| Gene Symbol: Gmfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031032.2 | NP_112294.1 | 4, 5 | 281 | ||
| GenBank mRNA | BC081778.1 | - | 4, 5 | 281 | 7192655 | |
| FQ222145.1 | - | 4, 5 | 343 | |||
| Z11558.1 | - | 3, 4 | 201 | |||