siRNA Details
| Gene Symbol: Musk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031061.1 | NP_112323.1 | 2 | 296 | ||
| GenBank mRNA | U34985.1 | - | 2 | 296 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Musk| Gene Symbol: Musk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031061.1 | NP_112323.1 | 2 | 296 | ||
| GenBank mRNA | U34985.1 | - | 2 | 296 | ||