siRNA Details
| Gene Symbol: Nup98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031074.2 | NP_112336.2 | 3, 4 | 366 | ||
| XM_006229892.2 | XP_006229954.1 | 3, 4 | 480 | |||
| XM_006229894.2 | XP_006229956.1 | 3, 4 | 328 | |||
| XM_006229895.2 | XP_006229957.1 | 3, 4 | 329 | |||
| GenBank mRNA | L39991.1 | - | 3, 4 | 338 | ||

Nup98
Genomic Map