siRNA Details
| Gene Symbol: SEC63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007214.4 | NP_009145.1 | 21 | 2746 | ||
| XM_011535399.2 | XP_011533701.1 | 21 | 2495 | |||
| XM_017010218.1 | XP_016865707.1 | 21 | 2575 | |||
| GenBank mRNA | AF119883.1 | - | 1 | 332 | ||
| AJ011779.1 | - | 21 | 2574 | |||
| AJ420532.1 | - | 3 | 1287 | |||
| AK027374.1 | - | 17 | 2019 | |||
| AK074835.1 | - | 21 | 2562 | |||
| AL137338.1 | - | 18 | 2093 | |||
| BC047221.1 | - | 21 | 2615 | 5277083 | ||
| BC048287.1 | - | 21 | 2602 | 5212007 | ||

SEC63
Genomic Map