siRNA Details
| Gene Symbol: Rbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031094.1 | NP_112356.1 | 6, 7 | 977 | ||
| XM_006255199.2 | XP_006255261.1 | 6, 7 | 1031 | |||
| XM_006255200.2 | XP_006255262.1 | 6, 7 | 1032 | |||
| XM_006255201.2 | XP_006255263.1 | 3, 4 | 1111 | |||
| GenBank mRNA | D55627.1 | - | 6, 7 | 977 | ||

Rbl2
Genomic Map