siRNA Details
| Gene Symbol: Rpl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031104.1 | NP_112366.1 | 5 | 399 | ||
| GenBank mRNA | BC058466.1 | - | 4 | 415 | 6887477 | |
| BC082750.1 | - | 4 | 390 | 7300771 | ||
| FQ222147.1 | - | 4 | 408 | |||
| FQ222660.1 | - | 4 | 408 | |||
| FQ222798.1 | - | 4 | 409 | |||
| FQ224693.1 | - | 4 | 407 | |||
| FQ228373.1 | - | 4 | 405 | |||
| X78444.1 | - | 5 | 399 | |||

Rpl22
Genomic Map