siRNA Details
Gene Symbol: Sctr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031115.1 | NP_112377.1 | 13 | 1699 | ||
GenBank mRNA | BC081781.1 | - | 13 | 1657 | 7189388 | |
X59132.1 | - | 13 | 1699 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
