siRNA Details
| Gene Symbol: Tgfb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031131.1 | NP_112393.1 | 3 | 837 | ||
| XM_006250448.2 | XP_006250510.1 | 2 | 1663 | |||
| GenBank mRNA | AF135598.1 | - | 2 | 447 | ||
| AF153012.2 | - | 3 | 837 | |||
| AF153013.1 | - | 2 | 753 | |||
| AJ132718.1 | - | 2 | 461 | |||

Tgfb2
Genomic Map