siRNA Details
| Gene Symbol: Tgfb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031131.1 | NP_112393.1 | 7 | 1417 | ||
| XM_006250448.2 | XP_006250510.1 | 6 | 2243 | |||
| GenBank mRNA | AF135598.1 | - | 6 | 1027 | ||
| AF153012.2 | - | 7 | 1417 | |||
| AF153013.1 | - | 6 | 1333 | |||

Tgfb2
Genomic Map