siRNA Details
| Gene Symbol: Dgkz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031143.1 | NP_112405.1 | 3 | 500 | ||
| XM_006234594.2 | XP_006234656.1 | 4 | 1172 | |||
| XM_006234596.2 | XP_006234658.1 | 3 | 335 | |||
| XM_008762037.1 | XP_008760259.1 | 3 | 2343 | |||
| GenBank mRNA | D78588.1 | - | 3 | 500 | ||

Dgkz
Genomic Map