siRNA Details
| Gene Symbol: Mdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031151.2 | NP_112413.2 | 2, 3 | 334 | ||
| GenBank mRNA | BC063165.1 | - | 2, 3 | 334 | 6918633 | |
| DQ402951.1 | - | 1, 2 | 168 | |||
| FQ213494.1 | - | 2, 3 | 287 | |||
| FQ213817.1 | - | 2, 3 | 251 | |||
| FQ215385.1 | - | 2, 3 | 294 | |||
| FQ215647.1 | - | 2, 3 | 295 | |||
| FQ216782.1 | - | 2, 3 | 325 | |||
| FQ217232.1 | - | 2, 3 | 286 | |||
| FQ224847.1 | - | 2, 3 | 231 | |||
| FQ224979.1 | - | 2, 3 | 313 | |||
| X04240.1 | - | 2, 3 | 275 | |||

Mdh2
Genomic Map