siRNA Details
| Gene Symbol: Fads2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031344.2 | NP_112634.1 | 2, 3 | 491 | ||
| XM_008760244.1 | XP_008758466.1 | 2, 3 | 2000 | |||
| GenBank mRNA | AB021980.1 | - | 2, 3 | 421 | ||
| BC081776.1 | - | 2, 3 | 491 | 7128630 | ||
| FQ210893.1 | - | 2, 3 | 420 | |||
| HQ909027.1 | - | 2, 3 | 325 | |||

Fads2
Genomic Map