siRNA Details
| Gene Symbol: Atrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031351.1 | NP_112641.1 | 2, 3 | 572 | ||
| GenBank mRNA | AB038387.1 | - | 2, 3 | 572 | ||
| AB038388.1 | - | 2, 3 | 572 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atrn| Gene Symbol: Atrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031351.1 | NP_112641.1 | 2, 3 | 572 | ||
| GenBank mRNA | AB038387.1 | - | 2, 3 | 572 | ||
| AB038388.1 | - | 2, 3 | 572 | |||