siRNA Details
Gene Symbol: Atrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031351.1 | NP_112641.1 | 3 | 602 | ||
GenBank mRNA | AB038387.1 | - | 3 | 602 | ||
AB038388.1 | - | 3 | 602 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031351.1 | NP_112641.1 | 3 | 602 | ||
GenBank mRNA | AB038387.1 | - | 3 | 602 | ||
AB038388.1 | - | 3 | 602 |