siRNA Details
Gene Symbol: Slc18a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031663.2 | NP_113851.1 | 1 | 2432 | ||
GenBank mRNA | U09211.1 | - | 1 | 2432 | ||
U09838.1 | - | 1 | 1689 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc18a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031663.2 | NP_113851.1 | 1 | 2432 | ||
GenBank mRNA | U09211.1 | - | 1 | 2432 | ||
U09838.1 | - | 1 | 1689 |