siRNA Details
| Gene Symbol: Stx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031665.2 | NP_113853.1 | 2 | 294 | ||
| GenBank mRNA | BC081769.1 | - | 2 | 294 | 7134641 | |
| U56815.1 | - | 2 | 270 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stx6| Gene Symbol: Stx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031665.2 | NP_113853.1 | 2 | 294 | ||
| GenBank mRNA | BC081769.1 | - | 2 | 294 | 7134641 | |
| U56815.1 | - | 2 | 270 | |||